Thermo Scientific 1120-1000 Storm Water Sample Bottle, Amber Glass; 1/Pk
Representative image only.
Thermo Scientific 1120-1000 Storm Water Sample Bottle, Amber Glass; 1/Pk
Thermo Scientific | Mfr: 1120-1000| Item: 9915902
Ideal for economical quarterly first-flush, EPA-compliant storm water grab samples
₹22,400.00INR / Each
₹22,400.00INR / Each
Product Description
Key Features
  • Collects a full one-liter grab sample within the first 30 minutes of a qualifying rain event
  • Mount in a variety of outfall types—storm grates, ditches, and streams
  • Disposable sampler requires no decontamination
More About this Item
The NALGENE Storm Water Sampler combines convenience and affordability to easily meet EPA requirements for storm water grab samples.
To collect the one-liter grab sample of first flush, simply position the sample in its protective mounting tube prior to the rain event, and leave it in place until after the storm. The sampler automatically collects the sample into the bottle and shuts off after the bottle is full, preventing dilution with later runoff. After the storm, the sampler can be retrieved at your convenience.
Complete instructions included and also available for download below. Brochure available for download to the left.
Mount disposable amber glass bottle with reusable Thermo Scientific Nalgene Sampler Mounting Kit 99159-12 at right.
EPA United States Environmental protection agency
Specifications
  • MaterialAmber Glass
  • Bottle TypeAmber Glass
  • DescriptionStorm Water Sample Bottle, Amber Glass; 1/Pk