Thermo Scientific 1010001 Conductivity Calibration Kit, NIST-Traceable Plug-in Resistor Set
Representative image only.
Thermo Scientific 1010001 Conductivity Calibration Kit, NIST-Traceable Plug-in Resistor Set
Thermo Scientific | Mfr: 1010001| Item: 1970446
Verify the performance of your Versa Star Pro and Star A conductivity meters
₹101,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹101,000.00INR / Each
Buy It Together
Current Item
Thermo Scientific 1010001 Conductivity Calibration Kit, NIST-Traceable Plug-in Resistor Set
Thermo Scientific 1010001 Conductivity Calibration Kit, NIST-Traceable Plug-in Resistor Set
1970446
₹101,000.00INR / Each
+ =
Total Price 125700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Six resistors simulate nominal conductance values between 1 µS and 100 mS
  • 8-pin Mini DIN connector
  • Traceable to NIST standard reference materials
More About this Item
The calibration kit contains six resistors that simulate nominal conductance values between 1 µS and 100 mS to verify the performance of Versa Star Pro and Orion 3-Star, 4-Star, and 5-Star meters.
Specifications
  • Compatible WithVersa Star Pro, 3-Star, 4-Star, and 5-Star meters
  • DescriptionConductivity Calibration Kit, NIST-Traceable Plug-In Resistor Set