Thermo Scientific 013010MD 4-Cell Conductivity Probe, Epoxy, K = 0.475, 8-Pin Mini DIN, 10-ft Cable
Representative image only.
Thermo Scientific 013010MD 4-Cell Conductivity Probe, Epoxy, K = 0.475, 8-Pin Mini DIN, 10-ft Cable
Thermo Scientific | Mfr: 013010MD| Item: 1970472
Take accurate conductivity measurements with these high-quality probes
₹96,130.00INR / Each
₹96,130.00INR / Each
Product Description
Key Features
  • Excellent durability and accuracy
  • Compatible with Star series meters
  • Epoxy body with graphite cells
  • 8-pin Mini DIN connector
  • 10-ft long cable
  • 1 year warranty
1 Year Warranty
Specifications
  • Cell ConstantK = 0.475
  • Body MaterialEpoxy
  • Use WithStar A meters
  • Cable Length (m)3
  • Cable Length (ft)10
  • Cell Configuration4-cell
  • Description4-Cell Conductivity Probe, Epoxy, K = 0.475, 8-Pin Mini DIN, 10-ft Cable