Thermal Transfer Compound, 4 oz Tube
Representative image only.
Thermal Transfer Compound, 4 oz Tube
| Item: 9378561
Increases thermal conductivity in thermo wells for faster response time
₹18,900.00INR / Each
₹18,900.00INR / Each
Product Description
Key Features
  • Designed to transfer heat away from heat-generating components
More About this Item
Silicone-free heat sink compound increases thermal conductivity in thermo wells for faster response time in comparison to air.
Specifications
  • DescriptionThermal Transfer Compound, 4 oz Tube