Texwipe TX716R Replacement Mop Heads, 6/cs
Representative image only.
Texwipe TX716R Replacement Mop Heads, 6/cs
Texwipe | Mfr: TX716R| Item: 8651822
Ideal choice for floor cleaning in controlled environments
₹147,000.00INR / Case of 6
₹147,000.00INR / Case of 6
Product Description
More About this Item
High-capacity, seamless, 100% knitted polyester head is constructed of tubular polyester for durability combined with low contamination characteristics. White, polyester-coated 52" steel handle is completely corrosion resistant. Quick open clamp encourages quick and easy mop head replacement. All components are autoclavable. Mop heads are cleanroom laundered and packaged. Order components separately below.
Specifications
  • DescriptionReplacement Mop Heads, 6/cs