Texwipe TX3342 Large Cleaning Validation Kit, 72 Areas
Representative image only.
Texwipe TX3342 Large Cleaning Validation Kit, 72 Areas
Texwipe | Mfr: TX3342| Item: 3367641
Securely transport swabs and vials with minimal contamination
₹142,000.00INR / Each
₹142,000.00INR / Each
Buy It Together
Current Item
Texwipe TX3342 Large Cleaning Validation Kit, 72 Areas
Texwipe TX3342 Large Cleaning Validation Kit, 72 Areas
3367641
₹142,000.00INR / Each
+ =
Total Price 150500.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Seventy-two vials, 144 swabs, and 72 labels.
Key Features
  • Kits packaged in cleanroom-compatible, polypropylene boxes
  • Doubled bagged to reduce risk of contamination
  • Each kit is individually lot coded
More About this Item
These total organic carbon (TOC) cleaning validation kits are designed to simplify sampling as part of a cleaning validation protocol. The kits offer you a convenient and secure method for transporting swab and vial samples from production to laboratory with minimal chance of contamination.
Packaged in cleanroom-compatible recyclable polypropylene boxes, the kits included 40 mL clear vials with bonded septa caps, low TOC Alpha® sampling swabs, and blank vial labels. Each box is double bagged for easier introduction into controlled or classified environments. Double-bagged boxes are packaged in a case liner forming the third layer of bag protection.
The pre-washed borosilicate glass vials have caps with a one-piece cap-and-septum design for easy extraction of vial content. The polyethylene over cap protects septum surface from contamination until time of analysis. Both the vials and caps are certified low TOC level (<10 ug/L, <10 ppb).
The swabs are designed with a double layered, double knit polyester head which allows maximum contaminant collection during sampling. Notched, break-away handle allows the head to be placed into the vial with minimal handling and contamination. The 100% polypropylene handle ensures no additional contaminants are introduced and provides excellent chemical resistance.
The labels are cleanroom compatible for vial sample identification and traceability.
Specifications
  • MaterialBorosilicate glass vials, double layered, double knit polyester head swabs with polypropylene handle
  • Handle Length (in)4
  • Swab Total Length (InchShort)5
  • Swab Total Length (cm)12.75
  • DescriptionLarge Cleaning Validation Kit, 72 Areas