Texwipe TX1065 Prewetted Cleanroom Wipes, 50 wipes/flat pack
Representative image only.
Texwipe TX1065 Prewetted Cleanroom Wipes, 50 wipes/flat pack
Texwipe | Mfr: TX1065| Item: 8643601
Save time and avoid solvent spillage with convenient presaturated wipe options
Call For Price
Call For Price
Product Description
Key Features
  • Save time and avoid solvent spillage with convenient presaturated wipe options
More About this Item
TechniSat® Wipes are nonwoven, 55% cellulose/45% polyester blend to combine the absorbent properties of a natural fiber with the strength and durability of a synthetic wiper. These economical wipes are saturated in 70% isopropyl alcohol/30% DI water—ideal for general purpose cleaning or in areas where nonvolatile residue (NVR) is a concern.
Note: Not available for sale to the state of California
Specifications
  • DescriptionPrewetted Cleanroom Wipes, 50 wipes/flat pack