Testo Flow Hood, 24" x 48"

Representative image only.

Testo Flow Hood, 24" x 48"

Testo | Mfr: 0554 4202| Item: 3000956

Improve performance of your airflow hood system

₹135,000.00INR / Each

₹135,000.00INR / Each
Product Description

Includes

Carrying case.

Key Features

  • Offers precision measurements at air inlets and outlets
  • Features an inner dimension of 23" x 47"
  • 2 year warranty

More About this Item

Choose these parts and accessories to meet your application needs.

Two year warranty
Specifications
  • DescriptionFlow Hood, 24" x 48"
Reviews
Q&A