Testo Flow Hood, 14" x 14"

Representative image only.

Testo Flow Hood, 14" x 14"

Testo | Mfr: 0554 4200| Item: 3000954

Improve performance of your airflow hood system

₹119,000.00INR / Each

₹119,000.00INR / Each
Product Description

Includes

Carrying case.

Key Features

  • Offers precision measurements at air inlets and outlets
  • Features an inner dimension of 13" x 13"
  • 2 year warranty

More About this Item

Choose these parts and accessories to meet your application needs.

Two year warranty
Specifications
  • DescriptionFlow Hood, 14" x 14"
Reviews
Q&A