Testo 16 Ft Silicone Hose for 512 Manometers
Representative image only.
Testo 16 Ft Silicone Hose for 512 Manometers
Testo | Mfr: 0554 0440| Item: 1038392
Flexible silicone tubing for obtaining differential measurement readings
₹25,300.00INR / Each
₹25,300.00INR / Each
Product Description
Key Features
  • 4-6 mm diameter
  • 700 hPa maximum pressure
  • Connects directly to Testo 512 Differential Manometers
  • 2 year warranty
Two year warranty
Specifications
  • Description16 Ft Silicone Hose for 512 Manometers 10383-92