TEL UK Air Flow Monitor for Bio Safety Cabinets, 0 to 2000 FPM Monitoring Range
Representative image only.
TEL UK Air Flow Monitor for Bio Safety Cabinets, 0 to 2000 FPM Monitoring Range
TEL UK | Mfr: M52BSC02US| Item: 3355670
Reliable, durable sensor element
₹227,000.00INR / Each
₹227,000.00INR / Each
Product Description
Includes
low voltage transformer and mounting bracket for surface mount. Call our Application Specialists if a flush mount requiring a 2" x 3" (5.1 x 7.6 cm) opening is preferred.
Key Features
  • Simple field calibration
  • Multiple alarms—audible, visual, and relay
More About this Item
Monitors maintain accurate response even when using low-flow hood conditions. Alarms activate based on sustained low-flow condition. Use pushbuttons to set alarm and perform easy two-point calibration. Monitors will also indicate high sash condition (requires proximity switch, not included) with configurable time delay. Record hood performance using RS-232 connection to computer.
Model 33556-50 features a horn alarm with green and red LED indicators. The graphic display model 33556-60 comes with an external velocity meter and shows current velocity in fpm with bar graph depiction of recent hood airflow performance; calibration instructions appear on screen. Choose model 33556-70 for its extra wide measurement range. It includes a remote velocity probe which can be installed in your ductwork for use with biosafety cabinets.
Specifications
  • Power (VAC)120
  • Power (Hz)60
  • DescriptionAir Flow Monitor for Bio Safety Cabinets, 0 to 2000 FPM Monitoring Range