Vendor supplied statement of traceability and three AA batteries.
Key Features
Stores up to 1000 measurements on the meter
±(0.04% of reading + 0.3°C) accuracy
Accepts thermocouple types J, K, T, E, N, R, S, and B
Ergonomically designed to minimize user fatigue
Easy-to-clean seamless case
SIngle-input datalogging thermometer with wireless capability
3 year warranty
More About this Item
Easily collect, store, and transmit temperature data for analysis or compliance reporting with these datalogging thermometers. Meters accurately record and store up to 1000 time-stamped measurements at the touch of a button on the meter; collect an unlimited amount of measurements by transferring them to a phone, PC, or cloud storage using Bluetooth® wireless technology. Simply download the Thermomoter Link™ mobile app for a two-way communication and control.
Both 931B and 932B models accept type J, K, T, E, N, R, S, and B thermocouple probes and have accuracy of ±(0.04% of reading + 0.3°C). Use the probe offset feature to zero out temperature probe errors, significantly improving overall measurement uncertainty. Increase productivity, efficiency, and quality using real-time statistical data. Thermometers display min, max, average, range, and standard deviation on demand, providing critical information and helping to analyze manufacturing process to make immediate adjustment for maximum efficiency and quality. Press the HOLD button to freeze display for reading later.
Easily view the readings on a large backlit LCD. The ergonomically designed thermometer allows for one-handed operation and minimizes user fatigue during extended use. The smooth, seamless case resists dirt and dust and is easy to clean. Thermometers provide up to 1000 hours of battery life using three AA batteries. Available in single- or dual-input model.
Specifications
Meter Measurement TypeThermocouple
Number Of Probe Channels1
Types Of Probes AcceptedTypes J, Type K, Type T, Type E, Type B, Type N, Type R, Type S
Min Temperature (° F)-418
Max Temperature (° F)3308
Min Temperature (° C)-250
Max Temperature (° C)1820
Accuracy±(0.04% of reading + 0.3°C)
Resolution<1000°: 0.1°>1000°: 1°
OutputBluetooth
Data LoggingYes
Memory Capacity1000 time-stamped measurements on the meter
Min/Max FunctionYes
Low Battery IndicatorYes
Display TypeBacklit LCD with auto resolution
Height (in)1 1/8
Width (in)3 1/4
Length (in)7 5/8
Battery Life1000 hours
Battery3 AA (included)
DescriptionSIngle-Input Datalogging Thermocouple Thermometer with Wireless Connectivity