Taylor 9878EPR Commercial Series Pen Style Waterproof Allergen Digital Thermometer with Large Head
Representative image only.
Taylor 9878EPR Commercial Series Pen Style Waterproof Allergen Digital Thermometer with Large Head
Taylor | Mfr: 9878EPR| Item: 9000152
Ideal for use in the food industry and all other general applications
Call For Price
for an additional
₹18,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
Taylor 9878EPR Commercial Series Pen Style Waterproof Allergen Digital Thermometer with Large Head
Taylor 9878EPR Commercial Series Pen Style Waterproof Allergen Digital Thermometer with Large Head
9000152
Call For Price
+ =
Total Price 18700.00 ADD 2 ITEMS TO CART
Product Description
Includes
storage sleeve with clip and 1 CR2032 battery.
Key Features
  • Maintain product safety with accurate readings
  • 1 year warranty
More About this Item
The real secret to great cooking is a Taylor thermometer. Thermometers are the only way to ensure that foods are cooked to the proper temperature, your oven is calibrated to the correct degree, and that your refrigerator is keeping your food cold and safe. Taylor is and has been the leader in food thermometers, with expertise over 150 years strong. You can count on Taylor for accuracy, superior performance, and dependability. This is why Taylor has tools chefs trust
Use in allergy programs and safety kits for peanuts, tree nuts, crustaceans, egg, milk, soy, and wheat
Easily read the digital display
Stainless steel construction is durable and NSF listed
1 Year Warranty
Specifications
  • Min Temperature (° F)-40
  • Max Temperature (° F)500
  • Min Temperature (° C)-40
  • Max Temperature (° C)260
  • Accuracy2%
  • Stem Length (in)4 1/2
  • Battery1 CR2023 (included)
  • DescriptionPen Style Waterproof Allergen Digital Thermometer with Large Head