Taylor 9867FDA Digital Pocket Thermometer with Folding Probe
Representative image only.
Taylor 9867FDA Digital Pocket Thermometer with Folding Probe
Taylor | Mfr: 9867FDA| Item: 9002576
1.5mm HACCP recommended probe diameter with Quick Tip Technology
₹15,800.00INR / Each
for an additional
₹18,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹15,800.00INR / Each
Buy It Together
Current Item
Taylor 9867FDA Digital Pocket Thermometer with Folding Probe
Taylor 9867FDA Digital Pocket Thermometer with Folding Probe
9002576
₹15,800.00INR / Each
+ =
Total Price 34500.00 ADD 2 ITEMS TO CART
Product Description
Includes
2 AAA batteries
Key Features
  • NSF certified
  • 4" Stainless Probe
  • Folding for storage
  • Accuracy of 0.5°F in HACCP temperatures ranges
  • Hold and automatic off functions
  • 1 year warranty
More About this Item
Taylor Precision Products has created a thermometer that has the commercial thermocouple, in a small folding package. The thermometer is easily stored as it just folds up to protect the 4" high quality stainless steel stem. This digital folding thermometer has a self set calibration system and has stabilized reading in less than 5 seconds. This Digital Precision thermometer has a 1.5mm FDA recommended probe diameter with Taylors Quick Tip Technology. Temperature read out runs from -40°F to 572°F / -40°C to 300°C and the resolution is 0.1°F / 0.1°C.
NSF international certified1 Year Warranty
Specifications
  • DescriptionTurbo Digital Pocket Thermocouple Thermometer