Taylor 9840PRN Commercial Series Pen Style Allergen Digital Thermometer
Representative image only.
Taylor 9840PRN Commercial Series Pen Style Allergen Digital Thermometer
Taylor | Mfr: 9840PRN| Item: 9000149
Ideal for use in the food industry and all other general applications
₹3,600.00INR / Each
for an additional
₹18,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹3,600.00INR / Each
Buy It Together
Current Item
Taylor 9840PRN Commercial Series Pen Style Allergen Digital Thermometer
Taylor 9840PRN Commercial Series Pen Style Allergen Digital Thermometer
9000149
₹3,600.00INR / Each
+ =
Total Price 22300.00 ADD 2 ITEMS TO CART
Product Description
Includes
storage sleeve with clip and 1 CR2032 battery.
Key Features
  • Maintain product safety with accurate readings
  • Temperature range -40 to 302°F (-40° to 150° C)
  • 3" LCD readout
  • step-down probe
  • on/off switch
  • 1 second updates
  • auto shut-off
  • 5" stainless steel stem
  • storage sleeve with clip
  • purple/allergy
  • 1.5 volt battery included
  • NSF
  • 1 year warranty
More About this Item
The real secret to great cooking is a Taylor thermometer. Thermometers are the only way to ensure that foods are cooked to the proper temperature, your oven is calibrated to the correct degree, and that your refrigerator is keeping your food cold and safe. Taylor is and has been the leader in food thermometers, with expertise over 150 years strong. You can count on Taylor for accuracy, superior performance, and dependability. This is why Taylor has tools chefs trust
Watch the temperature of your food with the Taylor 9840PRN purple digital pocket thermometer! This pocket thermometer's purple exterior is designed to remind your chefs to be careful about food allergens and prevent cross-contamination, and its digital display is large and easy to read. It comes with a protective sleeve to keep it safe when it's not in use, and its sensitive components can measure temperatures from -40 to 302 degrees Fahrenheit. And thanks to its On/Off switch, this thermometer gets the maximum life possible from its included 1.5V batter
1 Year Warranty
Specifications
  • Min Temperature (° F)-40
  • Max Temperature (° F)302
  • Min Temperature (° C)-40
  • Max Temperature (° C)150
  • Accuracy2%
  • Stem Length (in)5
  • Battery1 CR2032 (included)
  • DescriptionPen Style Allergen Digital Thermometer