Taylor 6092NBL Color-Coded Cross-Contamination Bimetal Pocket Thermometer - Blue
Representative image only.
Taylor 6092NBL Color-Coded Cross-Contamination Bimetal Pocket Thermometer - Blue
Taylor | Mfr: 6092NBLBC| Item: 9002570
Color Coded Thermometers Food Themometers
₹2,100.00INR / Each
for an additional
₹18,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹2,100.00INR / Each
Buy It Together
Current Item
Taylor 6092NBL Color-Coded Cross-Contamination Bimetal Pocket Thermometer - Blue
Taylor 6092NBL Color-Coded Cross-Contamination Bimetal Pocket Thermometer - Blue
9002570
₹2,100.00INR / Each
+ =
Total Price 20800.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 1" Diam. Dial and 5" stem
  • Shatter-proof lens and pocketed sleeve
  • 0°F to +220°F temperature range
  • NSF listed
  • Stainless steel construction
  • Five colors to optimal food safety
  • 1 year warranty
More About this Item
Color coded food safety pocket thermometer. This Taylor Precision 6092N, also known as a temperature indicator, is constructed of stainless steel. It has a 1" diameter dial and a 5" stem. This shatter-proof Taylor Precision 6092N has a pocketed sleeve. The sleeve features a unique built-calibration feature. This thermometer, Taylor Precision 6092N reads temperatures ranging from 0°F to +220°F. It is NSF listed. This Taylor Precision 6092N is available in five colors. Kitchens can use these pocket thermometers to color code. This food safe Taylor Precision 6092N helps to prevent cross-contamination. Purple is ideal for allergens, yellow for poultry, red for raw meat, brown for cooked meat and blue for fish.
1 Year Warranty
Specifications
  • Min Temperature (° F)0
  • Min Temperature (° C)-10
  • Max Temperature (° F)220
  • Max Temperature (° C)100
  • Division (° C)2
  • Division (° F)2
  • DescriptionCross Contamination 1" Bimetalal Pocket Thermometer - Blue