Taylor 5926 TruTemp Series Refrigerator / Freezer Analog Tube Thermometer with Safety Zones
Representative image only.
Taylor 5926 TruTemp Series Refrigerator / Freezer Analog Tube Thermometer with Safety Zones
Taylor | Mfr: 5926| Item: 9000126
Ensure proper temperature of your food
₹610.00INR / Each
for an additional
₹18,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹610.00INR / Each
Buy It Together
Current Item
Taylor 5926 TruTemp Series  Refrigerator / Freezer Analog Tube Thermometer with Safety Zones
Taylor 5926 TruTemp Series Refrigerator / Freezer Analog Tube Thermometer with Safety Zones
9000126
₹610.00INR / Each
+ =
Total Price 19310.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Compact size thermometer clips to rack or attaches with suction cup.
  • Temperature range -20 to 80°F (20 to 30°C).
  • Safe temperature zone indicators.
  • NSF listed
  • 1 year warranty
More About this Item
TruTemp Refrigerator / Freezer Glass Thermometer offers both a clip and a suction cup to accommodate any placement requirement. Dual scale shows Fahrenheit and Celsius, and includes a safe temperature zone guide. Non-toxic fill solution is safe to clean up in case of breakage.
1 Year WarrantyNSF international certified
Specifications
  • Min Temperature (° F)-20
  • Min Temperature (° C)-20
  • Max Temperature (° F)80
  • Max Temperature (° C)30
  • Accuracy2%
  • Dial Size (in)3
  • DescriptionRefrigerator / Freezer Analog Tube Thermometer with Safety Zones