Taylor 5921N Pro Series Oven / Grill Analog Thermometer with Zone References
Representative image only.
Taylor 5921N Pro Series Oven / Grill Analog Thermometer with Zone References
Taylor | Mfr: 5921N| Item: 9000145
Get more reliable, consistent results
₹3,300.00INR / Each
for an additional
₹18,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹3,300.00INR / Each
Buy It Together
Current Item
Taylor 5921N Pro Series Oven / Grill Analog Thermometer with Zone References
Taylor 5921N Pro Series Oven / Grill Analog Thermometer with Zone References
9000145
₹3,300.00INR / Each
+ =
Total Price 22000.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Large face with easy-to-read zone guide references.
  • Temperature range 100°F to 600°F.
  • Hangs or stands for secure placement in oven.
  • Durable stainless steel construction.
  • Dishwasher safe.
  • Measures 4 7/8" x 2 3/8
  • 1 year warranty
More About this Item
Pro Glass Oven Thermometer has a large face with easy-to-read temperature zone references, a durable stainless steel casing, and a mercury-free glass tube. Built-in hanger and stand works in any oven. Leave in oven to monitor temperatures. Dishwasher safe.
1 Year Warranty
Specifications
  • Min Temperature (° F)100
  • Max Temperature (° F)600
  • Max Temperature (° C)316
  • Division (° F)10
  • DescriptionOven / Grill Analog Thermometer with Zone References