Taylor 508 Connoisseur Pen Style Professional Digital Pocket Thermometer
Representative image only.
Taylor 508 Connoisseur Pen Style Professional Digital Pocket Thermometer
Taylor | Mfr: 508| Item: 9000123
Ideal for use in the food industry and all other general applications
₹4,100.00INR / Each
for an additional
₹18,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹4,100.00INR / Each
Buy It Together
Current Item
Taylor 508 Connoisseur Pen Style Professional Digital Pocket Thermometer
Taylor 508 Connoisseur Pen Style Professional Digital Pocket Thermometer
9000123
₹4,100.00INR / Each
+ =
Total Price 22800.00 ADD 2 ITEMS TO CART
Product Description
Includes
storage sleeve with clip and 1 CR2032 battery.
Key Features
  • Maintain product safety with accurate readings
  • Unique dot matrix display with animation for: on/off, F or C, selection hold
  • 3/8" digit height
  • Fast response stainless steel with silicone accents
  • Accurate to 446 deg. F
  • 1 year warranty
More About this Item
The real secret to great cooking is a Taylor thermometer. Thermometers are the only way to ensure that foods are cooked to the proper temperature, your oven is calibrated to the correct degree, and that your refrigerator is keeping your food cold and safe. Taylor is and has been the leader in food thermometers, with expertise over 150 years strong. You can count on Taylor for accuracy, superior performance, and dependability. This is why Taylor has tools chefs trust
The Taylor 508 Connoisseur Digital Instant Read Thermometer is used to ensure food has reached the proper temperature. The unique dot matrix display with 3/8" high digits shows the temperature while the auto off feature conserves battery power. The hold button stabilizes the temperature reading if desired and a switch allows the thermometer to display either Farenheit or Celsius. The Taylor 508 is constructed of stainless steel and the silicone grips are for safe and easy handling. The innovative shape and design ensure the thermometer will not roll off counters and a stem case is included with a convenient pocket clip.
1 Year Warranty
Specifications
  • Min Temperature (° F)-58
  • Max Temperature (° F)446
  • Min Temperature (° C)-50
  • Max Temperature (° C)230
  • DescriptionPen Style Professional Digital Pocket Thermometer