Taylor 3507 TruTemp Refrigerator / Freezer Analog Dial Thermometer with Safety Zones
Representative image only.
Taylor 3507 TruTemp Refrigerator / Freezer Analog Dial Thermometer with Safety Zones
Taylor | Mfr: 3507| Item: 9000125
Ensure proper temperature of your food
₹1,000.00INR / Each
for an additional
₹18,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹1,000.00INR / Each
Buy It Together
Current Item
Taylor 3507 TruTemp Refrigerator / Freezer Analog Dial Thermometer with Safety Zones
Taylor 3507 TruTemp Refrigerator / Freezer Analog Dial Thermometer with Safety Zones
9000125
₹1,000.00INR / Each
+ =
Total Price 19700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Large, 21⁄4-in. dial and red pointer are easy to read to quickly verify the temperature
  • Ideal for monitoring the temperature of refrigeration equipment, including walk-in coolers and freezers
  • Measures a temperature range of -20 to 80 degrees F
  • Color-coded dial indicates safe zones for refrigerator and freezer temperatures
  • Durable stainless steel construction resists corrosion, even in humid conditions
  • Can be hung on a shelf or sat standing on a flat surface
  • NSF Listed
  • 1 year warranty
More About this Item
TruTemp Refrigerator/Freezer Dial Thermometer has both a built-in hanger and stand. Compact dual-scale dial shows safe temperature zones. Durable stainless steel construction with a red pointer.
1 Year WarrantyNSF international certified
Specifications
  • Min Temperature (° F)-20
  • Min Temperature (° C)-20
  • Max Temperature (° F)80
  • Max Temperature (° C)30
  • Accuracy2%
  • DescriptionRefrigerator / Freezer Analog Dial Thermometer with Safety Zones