Taylor 3506 TruTemp Series Oven / Grill Analog Dial Thermometer with Dual-Scale
Representative image only.
Taylor 3506 TruTemp Series Oven / Grill Analog Dial Thermometer with Dual-Scale
Taylor | Mfr: 3506| Item: 9000143
Get more reliable, consistent results
₹1,200.00INR / Each
for an additional
₹18,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹1,200.00INR / Each
Buy It Together
Current Item
Taylor 3506 TruTemp Series Oven / Grill Analog Dial Thermometer with Dual-Scale
Taylor 3506 TruTemp Series Oven / Grill Analog Dial Thermometer with Dual-Scale
9000143
₹1,200.00INR / Each
+ =
Total Price 19900.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Easy-to-read 2 1/4" dial.
  • Temperture range 100°F to 600°F and 50°C to 300°C.
  • Bright blue and red dial graphics.
  • Red pointer.
  • Durable stainless steel.
  • Hangs or stands.
  • NSF listed.
  • 1 year warranty
More About this Item
TruTemp Bimetal Oven Thermometer features a compact dual-scale dial with a brightly colored guide to make reading easier and a durable stainless steel casing and glass lens. Built-in hanger and stand works in any oven. Leave in oven to monitor temperatures. Hand wash only.
1 Year Warranty
Specifications
  • Min Temperature (° F)100
  • Min Temperature (° C)50
  • Max Temperature (° F)600
  • Max Temperature (° C)316
  • Division (° C)5
  • Division (° F)5
  • Dial Size (in)2.25
  • DescriptionOven / Grill Analog Dial Thermometer with Dual-Scale