Taylor 1470N Classic Series Programmable Meat Thermometer with Timer and Alarm
Representative image only.
Taylor 1470N Classic Series Programmable Meat Thermometer with Timer and Alarm
Taylor | Mfr: 1470N| Item: 9000115
Digital probe thermometer is suitable for both hot and cold applications
₹5,400.00INR / Each
for an additional
₹18,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹5,400.00INR / Each
Buy It Together
Current Item
Taylor 1470N Classic Series Programmable Meat Thermometer with Timer and Alarm
Taylor 1470N Classic Series Programmable Meat Thermometer with Timer and Alarm
9000115
₹5,400.00INR / Each
+ =
Total Price 24100.00 ADD 2 ITEMS TO CART
Product Description
Includes
2 AAA batteries
Key Features
  • Classic Series Cooking Thermometer and Timer,
  • Temperature range 32° to 392°F (0° to 200° C)
  • Stainless Steel probe with 5 ft. silicone cord
  • On/Off switch
  • Ffolds down for storage with magnet on back for mounting unit to stove
  • 1 year warranty
More About this Item
This economic Taylor 1470N digital probe thermometer is suitable for both hot and cold applications. For extra user convenience, it has an alarm that sounds when the desired temperature is reached, and it can also be set to go off when a desired amount of time has passed. Its heat-resistant, 44" silicone cord and 6" stainless steel probe allow you to monitor cooking temperatures without opening the oven door. This Taylor probe thermometer has a range of 32-392 degrees Fahrenheit, 0-200 degrees Celsius. For best results, this thermometer is not recommended for use in oven temperatures greater than 392 degrees Fahrenheit. An adjustable, magnetic back allows you to conveniently place this thermometer on oven doors. Plus, its digital display is exceptionally easy to read!
1 Year Warranty
Specifications
  • Min Temperature (° F)32
  • Max Temperature (° F)392
  • Min Temperature (° C)0
  • Max Temperature (° C)200
  • Accuracy2%
  • AlarmSound
  • Battery2 AAA (included)
  • DescriptionProgrammable Meat Thermometer with Timer and Alarm