Taylor 1445 Pro Series Digital Fridge-Freezer Thermometer with Safety Zone
Representative image only.
Taylor 1445 Pro Series Digital Fridge-Freezer Thermometer with Safety Zone
Taylor | Mfr: 1445| Item: 9000132
Avoid food spoilage by maintaining adequate temperature
Call For Price
for an additional
₹18,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
Call For Price
Buy It Together
Current Item
Taylor 1445 Pro Series Digital Fridge-Freezer Thermometer with Safety Zone
Taylor 1445 Pro Series Digital Fridge-Freezer Thermometer with Safety Zone
9000132
Call For Price
+ =
Total Price 18700.00 ADD 2 ITEMS TO CART
Product Description
Includes
CR2032 lithium battery
CR2032 lithium battery
Key Features
  • Monitors the internal temperature of the fridge or freezer
  • Programmed with safety zone that activates timer if temperature goes outside the safe zone
  • “happy/sad” face indicator
  • Displays the amount of time that the temperature was not optimal
  • Can be displayed with magnet, clip or stand positioning
  • 1 year warranty
More About this Item
The Taylor Pro Digital Refrigerator and Freezer Thermometer ensures that the temperature in your appliance is cool enough to keep food fresh and safe to eat. This round thermometer can be placed in the fridge or freezer with either the attached magnet, clip or stand. It is programmed with a specific "safe" zone temperature that during power outages will record and display the amount of time the temperature was above optimal so you can monitor potential food safety concerns.
1 Year Warranty
Specifications
  • Min Temperature (° F)-4
  • Max Temperature (° F)140
  • Min Temperature (° C)-20
  • Max Temperature (° C)60
  • Accuracy2%
  • Memory CapacityMin/Max
  • AlarmHappy/Sad face "safe zone" temperature indicators
  • Mounting TypeClip, stand, or magnet
  • Display TypeDigital
  • Battery1 CR2032 (included)
  • DescriptionDigital Fridge-Freezer Thermometer with Safety Zone