Taylor 1442 Dual Zone Monitoring Thermometer with Alarms; -40 to 221F (-40 to 105C)
Representative image only.
Taylor 1442 Dual Zone Monitoring Thermometer with Alarms; -40 to 221F (-40 to 105C)
Taylor | Mfr: 1442| Item: 9446001
Dual channels, with separate and independent temperature measurement
₹12,400.00INR / Each
for an additional
₹18,700.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹12,400.00INR / Each
Buy It Together
Current Item
Taylor 1442 Dual Zone Monitoring Thermometer with Alarms; -40 to 221F (-40 to 105C)
Taylor 1442 Dual Zone Monitoring Thermometer with Alarms; -40 to 221F (-40 to 105C)
9446001
₹12,400.00INR / Each
+ =
Total Price 31100.00 ADD 2 ITEMS TO CART
Product Description
Includes
2 AAA batteries.
Key Features
  • Probe wires are 6 feet in length
  • Low-battery indication when replacement is necessary
  • 1 year warranty
More About this Item
This highly accurate dual channels instrument is designed for monitoring two separate and independent temperatures, ideal for use with temperature-critical materials. The thermometer has a weather resistant housing for indoor or outdoor use. Suitable for a variety of industrial and institutional applications, including manufacturing, food service (food storage), home and office. The thermometer displays temperature from each sensor probe in °C or °F, times (24 hour format only), and the status of each alarm event records. Each probe may be programmed with high and low limit alarms. The thermometer will capture in memory up to 60 alarm breeches per probe including time and duration of each breech. Alarm breeches will be retained in memory even if the battery dies. All operational fuctions can be password protected. The unit does not have an ON/OFF power button and is continually active once the batteries have been installed but unit does have an LCD idle function to preserve battery life.
1 Year Warranty
Specifications
  • Meter Measurement TypeDigital Indicator
  • Number Of Probe Channels2
  • Min Temperature (° F)-40
  • Max Temperature (° F)221
  • Min Temperature (° C)-40
  • Max Temperature (° C)105
  • Resolution0.1°
  • AlarmYes
  • Display Type0.8" Display
  • Height (in)3 1/2
  • Cable Length (in)72
  • Battery2 AAA (included)
  • DescriptionDual Zone Monitoring Thermometer with Alarms; -40 to 221F (-40 to 105C)