Supco 6" Circular Chart Recorder Paper, 6 Hour, -30 to 10°C, for Type J Thermocouple Recorders; 60/Pack
Representative image only.
Supco 6" Circular Chart Recorder Paper, 6 Hour, -30 to 10°C, for Type J Thermocouple Recorders; 60/Pack
Supco | Mfr: CR87-14| Item: 8005082
For Supco brand chart recorders
₹10,900.00INR / Pkg of 60
₹10,900.00INR / Pkg of 60
Buy It Together
Current Item
Supco 6" Circular Chart Recorder Paper, 6 Hour, -30 to 10°C, for Type J Thermocouple Recorders; 60/Pack
Supco 6" Circular Chart Recorder Paper, 6 Hour, -30 to 10°C, for Type J Thermocouple Recorders; 60/Pack
8005082
₹10,900.00INR / Pkg of 60
+ =
Total Price 182900.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Circular and strip charts
  • For CR87BC and CR87JC series recorders
  • 6 month warranty
More About this Item
Choose the paper compatible with your Supco chart recorder and application needs.
6 Month Warranty
Specifications
  • Chart TypeCircular
  • Chart Speed6 hour
  • Chart Diameter (in)6
  • Max Temperature (° F)50
  • Min Temperature (° F)-22
  • Max Temperature (° C)10
  • Min Temperature (° C)-30
  • Description6" Circular Chart Recorder Paper, 6 Hour, -30 to 10°C, for Type J Thermocouple Recorders; 60/Pack