Supco 6" Circular Chart Recorder Paper, 31 Day, 50 to 120°F; 60/Box

Representative image only.

Supco 6" Circular Chart Recorder Paper, 31 Day, 50 to 120°F; 60/Box

Supco | Mfr: CR87-21| Item: 8005263

For Supco brand chart recorders

₹11,200.00INR / Box of 60

₹11,200.00INR / Box of 60

Buy It Together

Current Item

Supco 6" Circular Chart Recorder Paper, 31 Day, 50 to 120°F; 60/Box
Supco 6" Circular Chart Recorder Paper, 31 Day, 50 to 120°F; 60/Box
8005263

₹11,200.00INR / Box of 60
+ =
Total Price 183200.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Circular and strip charts
  • For CR87B and CR87J series recorders
  • 1 year warranty

More About this Item

Choose the paper compatible with your Supco chart recorder and application needs.

1 Year Warranty
Specifications
  • Chart TypeCircular
  • Chart Speed31 day
  • Chart Diameter (in)6
  • Description6" Circular Chart Recorder Paper, 31 Day, 50 to 120°F; 60/Box
  • Warranty1 year
Reviews
Q&A