Stopper Hole Boring Set, Chrome-Plated Steel; 12 Borers/Set
Representative image only.
Stopper Hole Boring Set, Chrome-Plated Steel; 12 Borers/Set
| Item: 0629890
₹43,200.00INR / Set of 12
₹43,200.00INR / Set of 12
Product Description
Key Features
  • 6 month warranty
More About this Item
This chrome-plated steel cork borer set stays sharp. Flat blades permit easy boring of lightweight plastic, cork stoppers, and spongy articles.
Set consists of twelve borers with outside diameters of 4, 6, 7.5, 8.5, 10, 12, 13, 15, 17, 18.5, 19, and 20.5 mm.
6 Month Warranty
Specifications
  • MaterialSteel
  • DescriptionStopper Hole Boring Set, Chrome-Plated Steel; 12 Borers/Set