Spore Strips - 106 Population, 25 Strips/pack
Representative image only.
Spore Strips - 106 Population, 25 Strips/pack
| Item: 9470227
Call For Price
Call For Price
Product Description
More About this Item
Steam biological indicator: The spore strips can be used for sterility testing in various sterilization methods for both healthcare and industrial applications. For use in steam sterilization at 121degC to 135degC, low temperature steam with Formaldehyde and Chemiclave®. ETO biological indicator: The spore strips can be used for sterility testing in various sterilization for both healthcare and industrial
applications. For use in EtO sterilization at 600 mg/L. Gamma indicator: These are spore strips for validation of irradiation sterilization processes.
Specifications
  • DescriptionSpore Strips - 106 Population, 25 Strips/pack