Spill-X 78774 Multipurpose Spill Treatment/Clean Up Kit
Representative image only.
Spill-X 78774 Multipurpose Spill Treatment/Clean Up Kit
Spill-X | Mfr: 78774| Item: 8629208
Treat chemical spills in the lab or on the plant floor with a single step!
Call For Price
Call For Price
Buy It Together
Current Item
Spill-X 78774 Multipurpose Spill Treatment/Clean Up Kit
Spill-X 78774 Multipurpose Spill Treatment/Clean Up Kit
8629208
Call For Price
+
Spill-X 77255 Acid Agent Refill, 6 containers
8629250
Call For Price
+ + =
Total Price 0 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Spill Kit Treatment Guide, six polypropylene containers of agent, plastic case, one pair of rubber gloves and safety goggles, 100 pH test papers with color guide, scraper, two pick-up pans, and six disposal bags.
Key Features
  • Neutralizes hazardous liquid and chemical spills
  • Quick cleanups
  • 1 year warranty
More About this Item
These acid-neutralizing spill kits contain sorbents and other supplies which allow you to cleanup spills quickly. The easy-to-use shaker containers help neutralize hazardous liquid and chemical spills.
Order agent refills separately.
1 Year Warranty
Specifications
  • Chemical TreatedAcid/Caustic/Solvent
  • Agent ColorRed/Tan/Black
  • DescriptionMultipurpose Spill Treatment/Clean Up Kit