Spectra Por Tube-A-Lyzer Dialysis Device, 50000 MWCO, 8-10 mL; 3/Pk
Representative image only.
Spectra Por Tube-A-Lyzer Dialysis Device, 50000 MWCO, 8-10 mL; 3/Pk
Spectra Por | Mfr: 137009| Item: 0290654
Disposable dynamic device reduces dialysis times from 1 to 2 days down to 4 to 12 hours
₹53,900.00INR / Pkg of 3
₹53,900.00INR / Pkg of 3
Product Description
Key Features
  • Separate sample and flow-through buffer chambers significantly increase mass transfer rate and decrease run times
  • Design maintains maximum concentration gradient for up to 98% sample recovery, 99% sample purity and <10% sample dilution
  • 1 year warranty
More About this Item
This ready-to-use separation device delivers the efficiency of dynamic dialysis to significantly reduce dialysis from 1 to 2 days down to 4 to 12 hours. The Tube-A-Lyzer incorporates biotech-grade CE tubing that separates the sample chamber from the surrounding flow-through buffer chamber. The separate sample port provides easy access for sample loading, sample recovery and in-process testing.
The housing and end caps are polycarbonate (PC) with 3/16" (6 mm) barb inlet/outlet ports. Connect the housing to a buffer source and a waste collection vessel, or discharge directly to drain. Tube-A-Lyzer is non-pyrogenic and can be gamma sterilized (25 kGy). Dry packaged with 20% glycerin.
1 Year Warranty
Specifications
  • MaterialCellulose Ester, biotech-grade
  • Max Molecular Weight Cut Off (Mwco) (Dalton)50000
  • Max Sample Volume (mL)10
  • Min Sample Volume (mL)8
  • DescriptionDialysis Device, 50000 MWCO, 8-10 mL; 3/Pk