Spectra Por S/P 7 7 RC Dialysis Tubing, 15000 MWCO, 45mm/29mm; 16ft

Representative image only.

Spectra Por S/P 7 7 RC Dialysis Tubing, 15000 MWCO, 45mm/29mm; 16ft

Spectra Por | Mfr: 132124| Item: 0289058

Available in a variety of specific MWCO's

₹79,500.00INR / Box of 1

₹79,500.00INR / Box of 1

Buy It Together

Current Item

Spectra Por S/P 7 7 RC Dialysis Tubing, 15000 MWCO, 45mm/29mm; 16ft
Spectra Por S/P 7 7 RC Dialysis Tubing, 15000 MWCO, 45mm/29mm; 16ft
0289058

₹79,500.00INR / Box of 1
+ =
Total Price 115200.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Ideal for separations sensitive to heavy metal traces
  • 1 year warranty

More About this Item

For separations that require specific retentivity and are sensitive to sulfur and metals. EDTA-treated membranes contain no heavy metal traces or sulfur. Supplied wet in 0.1% sodium azide solution. Ready for use after rinsing and conditioning in dialyzing buffer. Each box contains 16-ft of membrane.

1 Year Warranty
Specifications
  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)15000
  • Flat Width (mm)45
  • Diameter (mm)29
  • Length Per Pack (ft)16
  • Diameter (in)1.142
  • Description7 RC Dialysis Tubing, 15000 MWCO, 45mm/29mm; 16ft
  • Warranty1 year
Reviews
Q&A