Spectra Por S/P 7 7 Membranes; MWCO, 1000; flat width, 18mm; diameter, 11.5mm

Representative image only.

Spectra Por S/P 7 7 Membranes; MWCO, 1000; flat width, 18mm; diameter, 11.5mm

Spectra Por | Mfr: 132103| Item: 0290254

Available in a variety of specific MWCO's

₹76,800.00INR / Box of 1

₹76,800.00INR / Box of 1

Buy It Together

Current Item

Spectra Por S/P 7 7 Membranes; MWCO, 1000; flat width, 18mm; diameter, 11.5mm
Spectra Por S/P 7 7 Membranes; MWCO, 1000; flat width, 18mm; diameter, 11.5mm
0290254

₹76,800.00INR / Box of 1
+ =
Total Price 109500.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Ideal for separations sensitive to heavy metal traces
  • 6 month warranty

More About this Item

For separations that require specific retentivity and are sensitive to sulfur and metals. EDTA-treated membranes contain no heavy metal traces or sulfur. Supplied wet in 0.1% sodium azide solution. Ready for use after rinsing and conditioning in dialyzing buffer. Each box contains 16-ft of membrane.

6 Month Warranty
Specifications
  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)1000
  • Flat Width (mm)18
  • Diameter (mm)11.5
  • Length Per Pack (ft)16
  • Diameter (in)0.453
  • Description7 Membranes; MWCO, 1000; flat width, 18mm; diameter, 11.5mm
  • Warranty6 month
Reviews
Q&A