Spectra Por S/P 7 7 Membranes; MWCO, 10,000; flath width, 12mm; diameter, 7.6mm

Representative image only.

Spectra Por S/P 7 7 Membranes; MWCO, 10,000; flath width, 12mm; diameter, 7.6mm

Spectra Por | Mfr: 132117| Item: 0290260

Available in a variety of specific MWCO's

₹76,800.00INR / Box of 1

₹76,800.00INR / Box of 1

Buy It Together

Current Item

Spectra Por S/P 7 7 Membranes; MWCO, 10,000; flath width, 12mm; diameter, 7.6mm
Spectra Por S/P 7 7 Membranes; MWCO, 10,000; flath width, 12mm; diameter, 7.6mm
0290260

₹76,800.00INR / Box of 1
+ =
Total Price 109500.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Ideal for separations sensitive to heavy metal traces
  • 6 month warranty

More About this Item

For separations that require specific retentivity and are sensitive to sulfur and metals. EDTA-treated membranes contain no heavy metal traces or sulfur. Supplied wet in 0.1% sodium azide solution. Ready for use after rinsing and conditioning in dialyzing buffer. Each box contains 16-ft of membrane.

6 Month Warranty
Specifications
  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)10000
  • Flat Width (mm)12
  • Diameter (mm)7.6
  • Length Per Pack (ft)16
  • Diameter (in)0.299
  • Description7 Membranes; MWCO, 10,000; flath width, 12mm; diameter, 7.6mm
  • Warranty6 month
Reviews
Q&A