Spectra Por S/P 2 Spectr/Por 2 Membranes Flat sheets; 240 x 240 mm; 25/pack

Representative image only.

Spectra Por S/P 2 Spectr/Por 2 Membranes Flat sheets; 240 x 240 mm; 25/pack

Spectra Por | Mfr: 132686| Item: 0290024

Available in a variety of dimensions to meet your applications needs

₹95,000.00INR / Pkg of 25

₹95,000.00INR / Pkg of 25
Product Description

Key Features

  • Regenerated Cellulose (RC) membrane for the gentle separation and purification of solutes
  • 6 month warranty

More About this Item

MWCO of 12,000 to 14,000 Dalton

6 Month Warranty
Specifications
  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)14000
  • Min Molecular Weight Cut Off (Mwco) (Dalton)12000
  • Square Sheet Length (mm)200
  • Square Sheet Width (mm)200
  • DescriptionSpectr/Por 2 Membranes Flat sheets; 240 x 240 mm; 25/pack
  • Warranty6 month
Reviews
Q&A