Spectra Por S/P 2 2 RC Dialysis Membrane Disc, 12000-14000 MWCO, 33mm Dia; 50/Pk
Representative image only.
Spectra Por S/P 2 2 RC Dialysis Membrane Disc, 12000-14000 MWCO, 33mm Dia; 50/Pk
Spectra Por | Mfr: 132482| Item: 0289064
Available in a variety of dimensions to meet your applications needs
₹60,200.00INR / Pkg of 50
₹60,200.00INR / Pkg of 50
Product Description
Key Features
  • Regenerated Cellulose (RC) membrane for the gentle separation and purification of solutes
  • 1 year warranty
More About this Item
MWCO of 12,000 to 14,000 Dalton
1 Year Warranty
Specifications
  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)14000
  • Min Molecular Weight Cut Off (Mwco) (Dalton)12000
  • Disc Diameter (mm)33
  • Description2 RC Dialysis Membrane Disc, 12000-14000 MWCO, 33mm Dia; 50/Pk