Spectra Por 4 RC Dialysis Tubing, 12-14 kD MWCO, 32mm Flat Width, 20.4mm dia; 100' L

Representative image only.

Spectra Por 4 RC Dialysis Tubing, 12-14 kD MWCO, 32mm Flat Width, 20.4mm dia; 100' L

Spectra Por | Mfr: 132703| Item: 0290043

Get gentle separation and purification of solutes

₹64,300.00INR / Box of 1

₹64,300.00INR / Box of 1
Product Description

Key Features

  • Available in a variety of diameters and flat widths
  • MWCO from 12,000 to 14,000 daltons
  • 6 month warranty

More About this Item

The RC Dialysis tubing is packaged dry to provide you with easy handling and storage. Available in 15 and 30 meter rolls to provide better economy for applications requiring larger amounts of tubing.
Most of the dry tubing rolls contain glycerin as a humectant which is easily removed by soaking 30 min in water.
Similar to SPECTRA/POR 2 tubing, except thicker. Dialysis time is slower, but at much less cost.

6 Month Warranty
Specifications
  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)14000
  • Min Molecular Weight Cut Off (Mwco) (Dalton)12000
  • Flat Width (mm)32
  • Diameter (mm)20.4
  • Length Per Pack (ft)100
  • Diameter (in)0.803
  • Description4 Membranes; flat width, 32mm; diameter, 20.4mm; length, 100'
  • Warranty6 month
Reviews
Q&A