Spectra Por 133336 Biotech-Grade RC Dialysis Tubing, 25000 MWCO, 10mm/6.4mm; 33ft

Representative image only.

Spectra Por 133336 Biotech-Grade RC Dialysis Tubing, 25000 MWCO, 10mm/6.4mm; 33ft

Spectra Por | Mfr: 133336| Item: 0289025

Ideal for concentration, purification, and desalination

₹78,600.00INR / Box of 1

₹78,600.00INR / Box of 1
Product Description

Key Features

  • High-purity membranes with no heavy metal or sulfide contaminants
  • Low protein adsoprtion
  • 1 year warranty

More About this Item

Regenerated cellulose (RC) offers broader temperature, pH, and chemical resistance; packaged as a 33-ft (10-m) roll, dry in 10% glycol

1 Year Warranty
Specifications
  • MaterialRegenerated Cellulose, biotech-grade
  • Max Molecular Weight Cut Off (Mwco) (Dalton)25000
  • Flat Width (mm)10
  • Diameter (mm)6.4
  • Length Per Pack (ft)33
  • Diameter (in)0.25
  • DescriptionBiotech-Grade RC Dialysis Tubing, 25000 MWCO, 10mm/6.4mm; 33ft
  • Warranty1 year
Reviews
Q&A