Spectra Por 132766 RC Dialysis Membrane Tubing Closure-Magnetic, 75MM Red 2/pk
Representative image only.
Spectra Por 132766 RC Dialysis Membrane Tubing Closure-Magnetic, 75MM Red 2/pk
Spectra Por | Mfr: 132766| Item: 0289929
The most convenient method for sealing dialysis tubing without the risk of leaking
₹25,900.00INR / Pkg of 2
₹25,900.00INR / Pkg of 2
Product Description
Key Features
  • Easy to open and reseal
  • 1 year warranty
More About this Item
Magnetic closures contain a fluorocarbon-coated magnetic bar, eliminating the need for conventional stir bars. This closure provides complete submersion and gentle rotation of the sample on a magnetic stirrer.
1 Year Warranty
Specifications
  • Tubing Sealing Width (mm)75
  • ColorRed
  • DescriptionRC Dialysis Membrane Tubing Closure-Magnetic, 75MM Red 2/pk