Spectra Por 132746 RC Dialysis Membrane Tubing Closure-Weighted, 75MM White 10/pk
Representative image only.
Spectra Por 132746 RC Dialysis Membrane Tubing Closure-Weighted, 75MM White 10/pk
Spectra Por | Mfr: 132746| Item: 0289927
The most convenient method for sealing dialysis tubing without the risk of leaking
₹37,400.00INR / Pkg of 10
₹37,400.00INR / Pkg of 10
Product Description
Key Features
  • Easy to open and reseal
  • 1 year warranty
More About this Item
Weighted closures feature a fluorocarbon-coated stainless steel bar. The weighted closure acts like an anchor to maintain a vertical orientation of the sample.
1 Year Warranty
Specifications
  • Tubing Sealing Width (mm)75
  • ColorWhite
  • DescriptionRC Dialysis Membrane Tubing Closure-Weighted, 75MM White 10/pk