Spectra Por 132745 Weighted Tubing Closures; max gripping width, 55mm; 10/box
Representative image only.
Spectra Por 132745 Weighted Tubing Closures; max gripping width, 55mm; 10/box
Spectra Por | Mfr: 132745| Item: 0289956
The most convenient method for sealing dialysis tubing without the risk of leaking
₹35,800.00INR / Box of 10
₹35,800.00INR / Box of 10
Product Description
Key Features
  • Easy to open and reseal
More About this Item
Weighted closures feature a fluorocarbon-coated stainless steel bar. The weighted closure acts like an anchor to maintain a vertical orientation of the sample.
Specifications
  • Tubing Sealing Width (mm)55
  • ColorWhite
  • DescriptionWeighted Tubing Closures; max gripping width, 55mm; 10/box