Spectra Por 132736 Plain tubing closures without weights; max gripping width, 35mm; 10/box
Representative image only.
Spectra Por 132736 Plain tubing closures without weights; max gripping width, 35mm; 10/box
Spectra Por | Mfr: 132736| Item: 0290050
The most convenient method for sealing dialysis tubing without the risk of leaking
₹31,500.00INR / Box of 10
₹31,500.00INR / Box of 10
Product Description
Key Features
  • Easy to open and reseal
  • 6 month warranty
More About this Item
Standard Closures are designed to float, providing buoyancy and vertical orientation when applied to the tubing top end. Autoclavable and available in several colors for easy sample identification.
6 Month Warranty
Specifications
  • Tubing Sealing Width (mm)35
  • ColorOrange
  • DescriptionPlain tubing closures without weights; max gripping width, 35mm; 10/box