SP Wilmad Caps for Disposable NMR Tubes, Assorted Color; 100/Pk
Representative image only.
SP Wilmad Caps for Disposable NMR Tubes, Assorted Color; 100/Pk
SP Wilmad | Mfr: 521-ASST-100| Item: 8399962
Easily track samples with different colored caps
₹4,200.00INR / Pkg of 100
₹4,200.00INR / Pkg of 100
Product Description
Key Features
  • Durable ethylene vinyl acetate construction
  • Compatible with 5 mm OD NMR tubes
More About this Item
These Wilmad-LabGlass standard 5 mm NMR tube caps come in a package of assorted colors. The different colors help track samples based on various reference solvents. These caps are compatible with 5 mm OD NMR tubes and are available in either an assortment pack of 100 or 1000 caps. When using chloroform or acetone as a reference solution, avoid direct contact between the sample and cap and long-term storage over two hours.
Specifications
  • SterileNo
  • Cap StyleDisposable for 5 mm OD NMR tubes
  • Cap MaterialEthylene Vinyl Acetate
  • Packaging100/Pk
  • DescriptionCaps for Disposable NMR Tubes, Assorted Color; 100/Pk