Soil Sampler Extension Rod, 26" Long
Representative image only.
Soil Sampler Extension Rod, 26" Long
| Item: 9902550
Available in 13 and 26-inch lengths, these rod extensions thread together in any combination
₹20,500.00INR / Each
₹20,500.00INR / Each
Product Description
More About this Item
Extensions are compatible with the following handles and sample tubes:
99025-04 Backsaver Handle
99025-06 Backsaver N-3 Handle
99025-02 Backsaver N-2 Handle
99025-60 T-Handle
99025-00 Conventional Handle
12-inch Wet or Dry Sampling Tubes
15-inch Wet or Dry Sampling Tubes
18-inch Wet or Dry Sampling Tubes
12-inch and 18-inch Large Diameter Sampling Tubes
12-inch and 18-inch Ag Consultant's Sampling Tubes
12-inch and 18-inch Zero Contamination Sampling Tubes
2-inch Concentric Sampling Tube
Specifications
  • Compatible WithSoil samplers 99025-XX
  • Length (InchShort)26
  • DescriptionSoil sampler extension rod, 26" long