Shimpo Funnel for Tachometers 25303-98 and 25304-93
Representative image only.
Shimpo Funnel for Tachometers 25303-98 and 25304-93
Shimpo | Mfr: FUNNEL-PT120| Item: 2530498
Designed specifically for PT-120 series tachometers
₹1,500.00INR / Each
₹1,500.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Measure rotation speed using a cone or funnel; cones are available in 1/4" or 1/2" diameter. Measure surface speed and distance with the measuring wheel.
Specifications
  • ColorBlack
  • Compatible WithTachometers 25303-98 and 25304-93
  • DescriptionFunnel for Tachometers 25303-98 and 25304-93