Sherwood Scientific Deproteinizing Concentrate for Flame Photometers, 6/pk
Representative image only.
Sherwood Scientific Deproteinizing Concentrate for Flame Photometers, 6/pk
Sherwood Scientific | Mfr: 00156182| Item: 0265531
Keep your flame photometer system running smothly
₹25,600.00INR / Pkg of 6
₹25,600.00INR / Pkg of 6
Product Description
More About this Item
These cleaning and diluting solutions are ideal for keeping your flame photometer in good working order,
If your samples have a high protein content, use this deproteinizing solution. Comes as six powder packets, each packet makes a 1000 mL solution.
Specifications
  • DescriptionDeproteinizing Concentrate for Flame Photometers, 6/pk