Sherwood Scientific Cleaning Solution for Flame Photometers, 500 mL
Representative image only.
Sherwood Scientific Cleaning Solution for Flame Photometers, 500 mL
Sherwood Scientific | Mfr: 00156184| Item: 0265579
Keep your flame photometer system running smothly
₹23,400.00INR / Each
₹23,400.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
These cleaning and diluting solutions are ideal for keeping your flame photometer in good working order,
After shutdown, rinse the photometer with general cleaning solution. Dilute the cleaning solution at a 1:100 ratio with deionized water before use.
1 Year Warranty
Specifications
  • DescriptionCleaning Solution for Flame Photometers, 500 mL