Sherwood Scientific 00156682 Tubing Disinfectant for Flame Photometers, 6 x 100 mL
Representative image only.
Sherwood Scientific 00156682 Tubing Disinfectant for Flame Photometers, 6 x 100 mL
Sherwood Scientific | Mfr: 00156682| Item: 0265581
Keep your flame photometer system running smothly
₹32,000.00INR / Pkg of 1
₹32,000.00INR / Pkg of 1
Product Description
Key Features
  • 1 year warranty
More About this Item
These cleaning and diluting solutions are ideal for keeping your flame photometer in good working order,
Use the tubing disinfectant when analyzing samples containing pathogenic organisms.
1 Year Warranty
Specifications
  • DescriptionTubing Disinfectant for Flame Photometers, 6 x 100 mL