Sherwood Scientific 00156681 Diluent Concentrate for Flame Photometers, 6 x 100 mL
Representative image only.
Sherwood Scientific 00156681 Diluent Concentrate for Flame Photometers, 6 x 100 mL
Sherwood Scientific | Mfr: 00156681| Item: 0265583
Keep your flame photometer system running smothly
₹36,800.00INR / Pkg of 6
₹36,800.00INR / Pkg of 6
Product Description
Key Features
  • 1 year warranty
More About this Item
These cleaning and diluting solutions are ideal for keeping your flame photometer in good working order,
Use the diluent concentrate for diluting samples and making blanks to zero your instrument; use at 1 part to 999 parts water.
1 Year Warranty
Specifications
  • DescriptionDiluent Concentrate for Flame Photometers, 6 x 100 mL