Sharpie SAN-13601 Industrial marker with fine tip, black
Representative image only.
Sharpie SAN-13601 Industrial marker with fine tip, black
Sharpie | Mfr: B00006IFEO| Item: 0996900
₹7,800.00INR / Pkg of 12
₹7,800.00INR / Pkg of 12
Product Description
More About this Item
Super-permanent black ink formulated for laboratory, industrial, and commercial use. Ink resists most chemical washes, steam, and heat up to 260°C (500°F).
Specifications
  • Tip StyleFine-point
  • Pen ColorBlack
  • DescriptionIndustrial marker with fine tip, black