Seward BA6040/STR Stomacher® 80 Biomaster Strainer Bags, Square Bottom; 200/PK
Representative image only.
Seward BA6040/STR Stomacher® 80 Biomaster Strainer Bags, Square Bottom; 200/PK
Seward | Mfr: BA6040/STR| Item: 3686000
Eliminates the risk of crosss contamination
Call For Price
Call For Price
Buy It Together
Current Item
Seward BA6040/STR Stomacher® 80 Biomaster Strainer Bags, Square Bottom; 200/PK
Seward BA6040/STR Stomacher® 80 Biomaster Strainer Bags, Square Bottom; 200/PK
3686000
Call For Price
+ +
Seward BA6090 Blending Bag Rack, Holds 10 Bags
3686002
Call For Price
=
Total Price 0 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • 1 year warranty
  • Manufactured from high-grade, food quality polythene
  • Filter / Strainer blender bags are for samples that produce a large amount of debris or fat or harder samples such as shellfish.
  • Filter / Strainer bags contain a robust strainer element to make it easier to pipette the sample produced (pore size 0.5mm, prevents pore blockage and non-homogenous sample).
  • Compatible with all 80 lab blenders, homogenizers
  • 1 year warranty
More About this Item
Choose from a variety of bags to use with your lab blender. Blending bags are made from high quality food grade nontoxic virgin LDPE granule, extruded to 65µm film thickness. All blender bags are irradiated sterile. A certificate or irridiation is available upon request.
The Stomacher 80 bag has been designed to fulfil all applications from blending, straining and storage including blender bags designed specifically for life science applications.
1 Year Warranty
Specifications
  • Bag TypePaddle Blender Bags
  • Bag Capacity (mL)80
  • Bag Capacity (L)0.08
  • DescriptionFilter Bags for Stomacher 80, square bottom with filter, 200 Per Pack